missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ NDSB-256, Calbiochem™,

Numéro de catalogue. 48001025GM
Modifier l'affichage
Click to view available options
Quantity:
25 g
Packaging:
Bouteille en verre
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
48001025GM 25 g Bouteille en verre
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 48001025GM Fournisseur MilliporeSigma™ Code fournisseur 48001025GM

Identifiants chimiques

CAS 81239-45-4
Molecular Formula C12H19NO3S
Molecular Weight (g/mol) 257.35
MDL Number MFCD00225018
InChI Key MEJASPJNLSQOAG-UHFFFAOYSA-N
Synonym Dimethylbenzylammonium Propane Sulfonate
PubChem CID 4386830
IUPAC Name 3-(benzyldiméthylazanyl)propane-1-sulfonate
SMILES C[N+](C)(CCCS([O-])(=O)=O)CC1=CC=CC=C1

Spécifications

Color Blanc
Quantity 25 g
Packaging Bouteille en verre
Percent Purity ≥99% (TLC)