missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Screw Top Vial Racks

Catalog No. 03389A
Change view
Click to view available options
Holds:
Vials
For Use With (Equipment):
4 mL Vials
Description:
Vial Rack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds For Use With (Equipment) Description
03389A Vials 4 mL Vials Vial Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03389A Supplier Thermo Scientific™ Supplier No. C401525

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Efficiently hold and organize vials while using them or for storage. Save space by stacking the 50 or 100 plastic vial racks on top of each other.

Store vials efficiently with Thermo Scientific™ National 4mL Screw Top Vial Racks, stackable for even more space savings.

  • Racks feature alphanumeric indexing for easier vial identification
  • Polypropylene vial racks are resistant to most solvents

Ordering Information
50 and 100 vial capacity racks available.

Specifications

Description Vial Rack
Color Clear
Holds Vials
Material Polypropylene
Positions 50
Shape Square
Stackable Yes
For Use With (Equipment) 4 mL Vials
Product Type Vial Rack

Limited warranty period:
Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.