missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nateglinide

Numéro de catalogue. AC462980010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC462980010 1 g
AC462980050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC462980010 Fournisseur Thermo Scientific Chemicals Code fournisseur 462980010
Il en reste null

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 105816-04-4
Molecular Formula C19H27NO3
Molecular Weight (g/mol) 317.43
InChI Key CZCVBLROPSHDRZ-YESZJQIVSA-N
IUPAC Name (2R)-3-phenyl-2-{[(1S,3S)-3-(propan-2-yl)cyclohexyl]formamido}propanoic acid
SMILES CC(C)[C@H]1CCC[C@@H](C1)C(=O)N[C@H](CC1=CC=CC=C1)C(O)=O

Spécifications

Melting Point 129.0°C to 130.0°C
Quantity 1 g
Formula Weight 317.42
Chemical Name or Material Nateglinide

RUO – Research Use Only