missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Narrow Mouth HDPE Translucent Reagent Bottles with Polypropylene Caps
Description
- Pack of 12 HDPE reagent bottles - Includes 12 narrow mouth 250 mL translucent HDPE bottles with polypropylene caps, designed for secure storage and handling of liquids in laboratory or industrial settings.
- Chemical resistance - Made from HDPE, offering excellent resistance to a variety of chemicals, making these bottles ideal for diverse lab and storage applications.
- Leak-proof tested - Rigorously tested to ensure leak protection during storage and transport. Features semi-buttress threads for a secure and reliable seal.
- Temperature range and design - Operates from -100°C to 120°C, with a 17.6 mm neck inner diameter for easy pouring and handling. Translucent design allows visibility for liquid monitoring.
- Versatile applications - Suitable for storing reagents, samples, and solutions. Commonly used in laboratories, industrial facilities, and research settings for safe liquid containment.
- Pack of 12, 250 mL narrow mouth reagent bottles made from HDPE with polypropylene caps.
- Designed for lab and industrial use, these non-sterile, leak-proof bottles are chemical resistant and operate between -100°C to 120°C.
- Ideal for storing reagents, samples, and solutions, the translucent design enhances liquid visibility for monitoring and measurement.
- Diameter 61.7 mm height 133 mm.
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Color | Translucent |
| Material | HDPE |
| No. per Pack | 12/Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Narrow |
| Autoclavable | Not Autoclavable |
| Capacity (English) | 8.45 oz. |
| Show More |