missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Narrow Mouth HDPE Translucent Reagent Bottles with Polypropylene Caps
Narrow mouth translucent HDPE bottles with polypropylene caps offers leakproof, chemical-resistant storage that is ideal for safely handling reagents, samples, and solutions in lab or industrial environments.
Specifications
| Color | Translucent |
|---|---|
| Material | HDPE |
| No. per Pack | 12 Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
S000075
|
Eisco Labs
CH5348PK12 |
4 mL |
N/A
|
|
|||||
|
S000076
|
Eisco Labs
CH5349PK12 |
8 mL |
N/A
|
|
|||||
|
S000077
|
Eisco Labs
CH5350PK12 |
15 mL |
N/A
|
|
|||||
|
S000078
|
Eisco Labs
CH5351PK12 |
30 mL |
N/A
|
|
|||||
|
S000079
|
Eisco Labs
CH5353PK12 |
125 mL |
N/A
|
|
|||||
|
S000080
|
Eisco Labs
CH5355PK12 |
500 mL |
N/A
|
|
|||||
|
S000081
|
Eisco Labs
CH5356PK12 |
1 L |
N/A
|
|
|||||
Description
- Pack Of 12 HDPE Reagent Bottles: Includes narrow mouth translucent HDPE bottles with polypropylene caps, designed for secure storage and handling of liquids in laboratory or industrial settings.
- Chemical Resistance: Made from HDPE, offering excellent resistance to a variety of chemicals, making these bottles ideal for diverse lab and storage applications.
- Leak-Proof Tested: Rigorously tested to ensure leak protection during storage and transport. Features semi-buttress threads for a secure and reliable seal.
- Temperature Range And Design: Operates from -100°C to +120°C. Translucent design allows visibility for liquid monitoring.
- Versatile Applications: Suitable for storing reagents, samples, and solutions. Commonly used in laboratories, industrial facilities, and research settings for safe liquid containment.
Specifications
| Translucent | |
| 12 Pk. | |
| Yes | |
| Narrow |
| HDPE | |
| Non-sterile | |
| No | |
| Reagent Bottle |