missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Narrow Mouth HDPE Translucent Reagent Bottles with Polypropylene Caps
Narrow mouth translucent HDPE bottles with polypropylene caps offers leakproof, chemical-resistant storage that is ideal for safely handling reagents, samples, and solutions in lab or industrial environments.
Supplier: Eisco Labs CH5350PK12
Description
- Pack Of 12 HDPE Reagent Bottles: Includes narrow mouth translucent HDPE bottles with polypropylene caps, designed for secure storage and handling of liquids in laboratory or industrial settings.
- Chemical Resistance: Made from HDPE, offering excellent resistance to a variety of chemicals, making these bottles ideal for diverse lab and storage applications.
- Leak-Proof Tested: Rigorously tested to ensure leak protection during storage and transport. Features semi-buttress threads for a secure and reliable seal.
- Temperature Range And Design: Operates from -100°C to +120°C. Translucent design allows visibility for liquid monitoring.
- Versatile Applications: Suitable for storing reagents, samples, and solutions. Commonly used in laboratories, industrial facilities, and research settings for safe liquid containment.
Chemical Identifiers
| Translucent | |
| 12 Pk. | |
| Yes | |
| Narrow |
| HDPE | |
| Non-sterile | |
| No | |
| Reagent Bottle |
Specifications
| 15 mL | |
| Translucent | |
| HDPE | |
| Non-sterile | |
| No | |
| 58 mm | |
| 25 mm |
| 0.5 oz. | |
| 14 mm | |
| 12 Pk. | |
| Yes | |
| Narrow | |
| Reagent Bottle | |
| Not Autoclavable |