missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Narrow Mouth HDPE Translucent Reagent Bottles with Polypropylene Caps

Catalog No. S000075
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
30 mL
125 mL
500 mL
1 L
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000075 4 mL
S000076 8 mL
S000077 15 mL
S000078 30 mL
S000079 125 mL
S000080 500 mL
S000081 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. S000075 Supplier Eisco Labs Supplier No. CH5348PK12

Narrow mouth translucent HDPE bottles with polypropylene caps offers leakproof, chemical-resistant storage that is ideal for safely handling reagents, samples, and solutions in lab or industrial environments.

  • Pack Of 12 HDPE Reagent Bottles: Includes narrow mouth translucent HDPE bottles with polypropylene caps, designed for secure storage and handling of liquids in laboratory or industrial settings.
  • Chemical Resistance: Made from HDPE, offering excellent resistance to a variety of chemicals, making these bottles ideal for diverse lab and storage applications.
  • Leak-Proof Tested: Rigorously tested to ensure leak protection during storage and transport. Features semi-buttress threads for a secure and reliable seal.
  • Temperature Range And Design: Operates from -100°C to +120°C. Translucent design allows visibility for liquid monitoring.
  • Versatile Applications: Suitable for storing reagents, samples, and solutions. Commonly used in laboratories, industrial facilities, and research settings for safe liquid containment.

Specifications

Capacity (Metric) 4 mL
Capacity (English) 0.13 oz.
Color Translucent
Diameter (Metric) Inner Neck 8.3 mm
Material HDPE
No. per Pack 12 Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 41 mm
Product Type Reagent Bottle
Diameter (Metric) 16 mm
Autoclavable Autoclavable
Show More Show Less