missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naphthol AS-BI bêta-D-Glucuronide 98,0+%, TCI America™
Spécifications
Spécifications
| Color | Blanc |
| Quantity | 25 mg |
| Formula Weight | 548.34 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | Naphthol AS-BI beta-D-Glucuronide [for Biochemical Research] |
Sécurité et manipulation
| TSCA | No |
| Recommended Storage | Congélateur |