missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene 98.0+%, TCI America™

Numéro de catalogue. p-7134011
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
N088525G 25 g
N0885500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. N088525G Fournisseur TCI America Code fournisseur N088525G
Il en reste null

Identifiants chimiques

CAS 91-20-3
Molecular Formula C10H8
Molecular Weight (g/mol) 128.17
MDL Number MFCD00001742
InChI Key UFWIBTONFRDIAS-UHFFFAOYSA-N
Synonym naphthalin, naphthene, camphor tar, tar camphor, white tar, albocarbon, naphthaline, moth flakes, moth balls, naphtalene
PubChem CID 931
ChEBI CHEBI:16482
IUPAC Name naphthalene
SMILES C1=CC2=CC=CC=C2C=C1

Spécifications

Melting Point 81°C
Color White
Boiling Point 218°C
Quantity 25 g
UN Number 1334
Formula Weight 128.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Naphthalene
EINECSNumber (4)-0311
RTECSNumber QJ0525000
TSCA Yes