missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene, Analytical Standard, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020686
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020686 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020686 Supplier Merck Emd Millipore Supplier No. 84679250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 91-20-3
Vapor Pressure 0.03 mmHg (25°C)
Vapor Density 4.4 (vs. air)
Physical Form Neat
Linear Formula C10H8
MDL Number MFCD00001742
Melting Point 79.5°C to 81.0°C; 80°C to 82°C
Boiling Point 218°C
Quantity 250 mg
Formula Weight 128.17
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less