missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene, 99%, Spectrum™ Chemical

Catalog No. 18611904
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611904 500 g Poly Pail
18611902 2.5 kg Poly Pail
18611901 12 kg Poly Pail
18611903 50 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611904 Supplier Spectrum Chemical Mfg Cor Supplier No. N1005500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91-20-3

Specifications

CAS Max % 1
Boiling Point 220°C/428°F
Packaging Poly Pail
Quantity 500 g
Percent Purity 0.99
Grade Ungraded
Physical Form Crystals or Flakes
Chemical Name or Material Naphthalene

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.