missing translation for 'onlineSavingsMsg'
Learn More

Naphazoline hydrochloride, 99%

Catalog No. AC179660250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC179660250 25 g Plastic bottle
AC179661000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC179660250 Supplier Thermo Scientific Chemicals Supplier No. 179660250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 550-99-2 | C14H14N2·ClH | 246.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 550-99-2
Molecular Formula C14H14N2·ClH
Molecular Weight (g/mol) 246.74
MDL Number MFCD00012554
InChI Key DJDFFEBSKJCGHC-UHFFFAOYSA-N
Synonym naphazoline hydrochloride, albalon, naphazoline hcl, rhinantin, rhinoperd, stricylon, naphcon, niazol, rinofug, vasocon
PubChem CID 11079
ChEBI CHEBI:7470
IUPAC Name 2-(naphthalen-1-ylmethyl)-4,5-dihydro-1H-imidazole;hydrochloride
SMILES C1CN=C(N1)CC2=CC=CC3=CC=CC=C32.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 254.0°C to 260.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 25 g
Merck Index 15, 6453
Solubility Information Solubility in water: 170g/L (20°C).
Formula Weight 246.74
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Naphazoline hydrochloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
Do not eat,drink or smoke when using this product.
Obtain special instructions before use.
EINECSNumber 208-989-2
RTECSNumber NJ4375000
TSCA TSCA

RUO – Research Use Only