missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Sodium Hydroxide (NaOH) Solutions, 0.1–1.0 M
GREENER_CHOICE

Catalog No. A4782601
Change view
Click to view available options
Quantity:
20 L
200 L
Concentration:
0.1 M
0.5 M
1.0 M
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
A4782601 20 L 0.1 M
A4782602 200 L 0.1 M
A4782801 20 L 0.5 M
A4782802 200 L 0.5 M
A4782901 20 L 1.0 M
A4782902 200 L 1.0 M
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. A4782601 Supplier Gibco™ Supplier No. A4782601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gibco NaOH solutions offer more options for supply security and dual sourcing of the highest volume process liquid in bioprocessing manufacturing.

Gibco NaOH solutions offer more options for supply security and dual sourcing of the highest volume process liquid in bioprocessing manufacturing. Available in three different grades and two off-the-shelf sizes, these solutions provide outsourcing efficiencies to help reclaim time, space, and other core resources.

Features of Gibco NaOH solutions include:

  • 0.1 M, 0.5 M, and 1 M concentrations
  • Multi-compendial grade raw materials
  • cGMP manufacturing to 21CFR820
  • 20 L and 200 L ready-to-use single-use bioprocessing containers

Flexibility

Gibco NaOH solutions provide outsourcing efficiencies for customers manufacturing internally but seeking to partially outsource, as well as for customers with batch sizes that do not justify custom ordering and customers already outsourcing to Thermo Fisher Scientific but seeking additional flexibility. They are produced in the most commonly used concentrations and single-use containers.

Applications of Gibco NaOH solutions

  • pH adjustment
  • Facility/equipment sanitization
  • Chromatography column cleaning, sanitization and storage

Specifications

Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2
Melting Point 0°C
Boiling Point 100°C
Color Clear
Concentration 0.1 M
pH ≥12 to ≤14
Quantity 20 L
Molecular Formula NaOH
Packaging Bioprocess Container
Synonym Caustic soda, Acidity regulatory, Lye or Soda lye
Molecular Weight (g/mol) 40
Percent Purity 95 to 100%
Grade Compendial
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.