missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific NanoDrop UltraC FL Security and IQOQ Bundle

Catalog No. 13400543
Change view
Click to view available options
Spectral Bandwidth:
≤1.8 nm (FWHM at Hg 254 nm)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Spectral Bandwidth
13400543 ≤1.8 nm (FWHM at Hg 254 nm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13400543 Supplier Thermo Scientific Supplier No. NDULTRACFLSECUS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Delivers fast, accurate microvolume UV-Vis and fluorescence analysis with SciVault 2 for 21 CFR Part 11 and IQOQ compliance.

  • The NanoDrop™ Ultra Spectrophotometers and Fluorometers represent the next generation of microvolume detection, offering exceptional accuracy and sensitivity for nucleic acid and protein analysis using as little as 1.0 μL.
  • Designed to integrate seamlessly into workflows across genomics, proteomics, molecular biology, biotechnology, pharmaceutical, and life sciences research applications.
  • Provides unmatched precision, efficiency, and reliability in a compact design for consistent microvolume analysis.
  • Each bundle includes one NanoDrop Ultra instrument with Thermo Scientific™ SciVault™ 2 activation and a model-specific IQ/OQ kit for qualification and verification.
  • Incorporates Thermo Scientific™ Acclaro™ Sample Intelligence technology for improved measurement accuracy and contaminant detection.
  • Quantify and qualify DNA, RNA, and protein samples in seconds using only 1 to 2 μL, with full spectral data available for downstream analysis decisions.
  • Includes an optional cuvette position to provide additional flexibility for a variety of experimental workflows.
  • Thermo Scientific™ SciVault™ 2 software equips laboratories with the tools required to comply with US FDA 21 CFR Part 11 data integrity regulations.
  • Integrates with NanoDrop Ultra software to manage user accounts, digital signatures, and audit trails, simplifying compliance workflows.
  • Supports both on-board user account management and domain-based access using Windows authentication for flexible control.
  • Centralized deployment enables privilege control and audit log management across multiple NanoDrop Ultra instruments in distributed labs.
  • Fully integrated qPCR recipe calculator uses built-in or custom libraries to automate reaction setup and calculations based on sample concentration.
  • Auto-Measure and Auto-Blank functions streamline workflows by capturing instant measurements when the pedestal arm is lowered; can be toggled ON or OFF via the touchscreen interface.
  • Enables direct export of data to cloud storage platforms, including Microsoft OneDrive, Google Drive, and Thermo Fisher Connect.
  • Facilitates direct LIMS integration with an integrated Restful API for seamless data management and reporting.
  • Offers flexible data-to-printer options via Wi-Fi, Ethernet, or USB, or print from a PC using NanoDrop Ultra PC Control software.
  • Allows full customization and preview of exported data files to meet downstream documentation or compliance needs.
  • Model-specific IQ/OQ Kits include a USB flash drive with IQ/OQ procedures, CHEM-PR1-KIT, CHEM-PV-1, and Cuvette Qualification Kit (840-257300) if a cuvette model is purchased, or CHEM-FL-1 for fluorescence models.
  • For research use only; not for diagnostic purposes.

Specifications

Detector Type 2048-element CMOS Linear Image Sensor
Display Color Touchscreen
Languages English, Chinese, French, Japanese, German, Spanish, Polish, Italian, Korean
Light Source Xenon Flash, LED
Product Line NanoDrop
Spectral Bandwidth ≤1.8 nm (FWHM at Hg 254 nm)
Type Spectrophotometers Bundle
Voltage 100/240 VAC
Wavelength Range 190 to 850 nm
Wavelength Selection Automatic