Learn More
Therapak™ NanoCool™ Refrigerated Specimen Shipper
Description
Comprend
Sturdy transport box.
- Portable unit maintains temperature from 2-8°C — ideal for overnight or international shipments
- Weighs less and requires less space than conventional polystyrene boxes and ice packs, reducing shipping costs
- Vacuum Insulated Panel (VIP) compartment transfers heat by evaporating small quantities of water at low pressure; a heat absorption method that's seven times more efficient than ice
- Separate NanoCool cooler fits atop the VIP compartment and can be activated at room temperature with the push of a button
- Small shipper holds up to 18 standard (13 × 100mm) transport tubes; large shipper holds up to 36 tubes
Order Info
One-time use only; cannot be reconditioned
Spécifications
Spécifications
| Quantity | Up to 18 standard 13 x 100mm tubes |
| Includes | Transport box |
| Type | Portable Unit |
| Duration | For overnight or international shipping |