missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-(tert-Butoxycarbonyl)-L-asparagine 98.0+%, TCI America™

Catalog No. B162710G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B162710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B162710G Supplier TCI America Supplier No. B162710G
Only null left

Chemical Identifiers

CAS 7536-55-2
Molecular Formula C9H16N2O5
Molecular Weight (g/mol) 232.236
MDL Number MFCD00038152
InChI Key FYYSQDHBALBGHX-YFKPBYRVSA-N
Synonym boc-asn-oh, boc-l-asparagine, boc-asn, n-tert-butoxycarbonyl-l-asparagine, tert-butoxycarbonylasparagine, nalpha-tert-butoxycarbonyl-l-asparagine, boc-asparagine, tert-butoxycarbonyl-l-asparagine, n-tert-butoxycarbonyl asparagine, tert-butyloxycarbonyl-l-asparagine
PubChem CID 82035
ChEBI CHEBI:3146
IUPAC Name (2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O

Specifications

Melting Point 166°C
Color White
Quantity 10 g
Formula Weight 232.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(tert-Butoxycarbonyl)-L-asparagine