missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-(tert-Butoxycarbonyl)-D-arginine Hydrochloride Monohydrate 98.0+%, TCI America™

Catalog No. B22045G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22045G 5 g
B22041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22045G Supplier TCI America Supplier No. B22045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 114622-81-0
Molecular Formula C11H25ClN4O5
Molecular Weight (g/mol) 328.794
MDL Number MFCD00065554
InChI Key OVXLPYFDJUFEHQ-KLXURFKVSA-N
Synonym boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, boc-arg-oh.hcl.h2o, boc-d-arg-oh hcl h2o, boc-arg-oh hcl h2o, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, a-boc-d-arginine hydrochloride monohydrate, pubchem14932, boc-l-arg-ohhclh2o
PubChem CID 2729004
IUPAC Name (2S)-5-(diaminomethylideneamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate;hydrochloride
SMILES CC(C)(C)OC(=O)NC(CCCN=C(N)N)C(=O)O.O.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 310.78
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(tert-Butoxycarbonyl)-D-arginine Hydrochloride Monohydrate