missing translation for 'onlineSavingsMsg'
Learn More

N-alpha,N-epsilon-Bis[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysine 98.0+%, TCI America™

Catalog No. B49315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49315G 5 g
B493125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49315G Supplier TCI America Supplier No. B49315G
Only null left

Chemical Identifiers

CAS 78081-87-5
Molecular Formula C36H34N2O6
Molecular Weight (g/mol) 590.676
MDL Number MFCD00080275
InChI Key BMJRTKDVFXYEFS-XIFFEERXSA-N
Synonym fmoc-lys fmoc-oh, n,n'-bis-fmoc-l-lysine, fmoc-l-lys fmoc-oh, n, a,n, a-di-fmoc-l-lysine, 2s-2,6-bis 9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, fmoclys fmoc oh, pubchem18973, n, n'-bis-fmoc-l-lysine, n-a,n-epsilon-di-fmoc-l-lysine, nalpha,nepsilon-di-fmoc-l-lysine
PubChem CID 13783708
IUPAC Name (2S)-2,6-bis(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46

Specifications

Melting Point 135°C
Color White
Quantity 5 g
Formula Weight 590.68
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-alpha,N-epsilon-Bis[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysine
TSCA No
Recommended Storage Refrigerator