missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Fmoc-Ngamma-trityl-L-asparagine, 97%
CAS: 132388-59-1 | C38H31N2O5 | 595.68 g/mol
Supplier: Thermo Scientific Chemicals H5953614
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Nalpha-Fmoc-Ngamma-trityl-L-asparagine | |
| 201°C to 204°C | |
| MFCD00077049 | |
| 4343823 | |
| KJYAFJQCGPUXJY-UMSFTDKQSA-M | |
| [O-]C(=O)[C@H](CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 595.68 | |
| 596.67 |
| 132388-59-1 | |
| C38H31N2O5 | |
| 25 g | |
| fmoc-asn trt-oh, nalpha-fmoc-ngamma-trityl-l-asparagine, ambotzfaa1015, n-fmoc-n4-trityl-l-asparagine, pubchem9947, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-oxo-4-tritylamino butanoic acid, fmoc-n'-trityl-l-asparagine, nalpha-9h-fluoren-9-ylmethoxy carbonyl-ngamma-trityl-l-asparagine, n-9-fluorenyl methoxy carbonyl-n-trityl-l-asparagine | |
| −15° (c=1 in Methanol) | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-(tritylamino)butanoic acid | |
| 640248 | |
| 97% |
Chemical Identifiers
| 132388-59-1 | |
| 595.68 | |
| KJYAFJQCGPUXJY-UMSFTDKQSA-M | |
| 640248 | |
| [O-]C(=O)[C@H](CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
| C38H31N2O5 | |
| MFCD00077049 | |
| fmoc-asn trt-oh, nalpha-fmoc-ngamma-trityl-l-asparagine, ambotzfaa1015, n-fmoc-n4-trityl-l-asparagine, pubchem9947, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-oxo-4-tritylamino butanoic acid, fmoc-n'-trityl-l-asparagine, nalpha-9h-fluoren-9-ylmethoxy carbonyl-ngamma-trityl-l-asparagine, n-9-fluorenyl methoxy carbonyl-n-trityl-l-asparagine | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-(tritylamino)butanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only