missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-Ngamma-trityl-L-asparagine, 97%

Catalog No. p-7039984
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5953606 5 g
AAH5953614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5953606 Supplier Thermo Scientific Chemicals Supplier No. H5953606
Only null left

CAS: 132388-59-1 | C38H31N2O5 | 595.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132388-59-1
Molecular Formula C38H31N2O5
Molecular Weight (g/mol) 595.68
MDL Number MFCD00077049
InChI Key KJYAFJQCGPUXJY-UMSFTDKQSA-M
Synonym fmoc-asn trt-oh, nalpha-fmoc-ngamma-trityl-l-asparagine, ambotzfaa1015, n-fmoc-n4-trityl-l-asparagine, pubchem9947, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-oxo-4-tritylamino butanoic acid, fmoc-n'-trityl-l-asparagine, nalpha-9h-fluoren-9-ylmethoxy carbonyl-ngamma-trityl-l-asparagine, n-9-fluorenyl methoxy carbonyl-n-trityl-l-asparagine
PubChem CID 640248
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-(tritylamino)butanoic acid
SMILES [O-]C(=O)[C@H](CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 201°C to 204°C
Quantity 5 g
Beilstein 4343823
Optical Rotation −15° (c=1 in Methanol)
Formula Weight 596.67
Percent Purity 97%
Chemical Name or Material Nalpha-Fmoc-Ngamma-trityl-L-asparagine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only