missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-Nepsilon-carbamoyl-L-lysine, 95%

Catalog No. AAH6618903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6618903 1 g
AAH6618906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6618903 Supplier Thermo Scientific Chemicals Supplier No. H6618903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 201485-17-8 | C22H25N3O5 | 411.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 201485-17-8
Molecular Formula C22H25N3O5
Molecular Weight (g/mol) 411.46
MDL Number MFCD00237387
InChI Key VJZUCXPETVPQIB-TWYLJJHKNA-N
Synonym fmoc-hocit-oh, fmoc-l-homocitrulline, fmoc-hcit-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-ureidohexanoic acid, fmoc-homocit-oh, 2s-6-carbamoylamino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzfaa1383, fmoc-lys carbamoyl-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-homocitrulline, s-2-9h-fluoren-9-yl methoxy carbonyl-amino-6-ureidohexanoic acid
PubChem CID 7019813
IUPAC Name (2S)-6-(carbamoylamino)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
SMILES NC(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Quantity 1 g
Formula Weight 411.46
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Nalpha-Fmoc-Nepsilon-carbamoyl-L-lysine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only