missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Carbobenzoxy-D-tryptophan 98.0+%, TCI America™

Catalog No. C21305G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21305G 5 g
C21301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21305G Supplier TCI America Supplier No. C21305G
Only null left

Chemical Identifiers

CAS 2279-15-4
Molecular Formula C19H18N2O4
Molecular Weight (g/mol) 338.36
MDL Number MFCD00037945
InChI Key AHYFYYVVAXRMKB-QGZVFWFLSA-N
Synonym z-d-trp-oh, n-cbz-d-tryptophan, z-d-tryptophan, cbz-d-trp-oh, nalpha-cbz-d-tryptophan, r-2-benzyloxy carbonyl amino-3-1h-indol-3-yl propanoic acid, n a-benzyloxycarbonyl-d-tryptophan, chembl65134, nalpha-carbobenzoxy-d-tryptophan, d-tryptophan, n-phenylmethoxy carbonyl
PubChem CID 700663
IUPAC Name (2R)-2-{[(benzyloxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoic acid
SMILES OC(=O)[C@@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 5 g
Formula Weight 338.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Carbobenzoxy-D-tryptophan