missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-Nepsilon-Fmoc-L-lysine, 98%

Catalog No. AAH6112614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6112614 25 g
AAH6112606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6112614 Supplier Thermo Scientific Chemicals Supplier No. H6112614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 84624-27-1 | C26H32N2O6 | 468.55 g/mol

Nalpha-Boc-Nepsilon-Fmoc-L-lysine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha-Boc-Nepsilon-Fmoc-L-lysine is used as a pharmaceutical intermediate.

Solubility
Soluble in dimethylformamide. Insoluble in water.

Notes
Keep container tightly sealed. Store refrigerated conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 84624-27-1
Molecular Formula C26H32N2O6
Molecular Weight (g/mol) 468.55
MDL Number MFCD00062032
InChI Key JYEVQYFWINBXJU-ANBDAQEENA-N
Synonym boc-lys fmoc-oh, n-boc-n'-fmoc-l-lysine, n2-boc-n6-fmoc-l-lysine, n-boc-n-fmoc-l-lysine, boclys fmoc, n-epsilon-fmoc-alpha-t-boc-l-lysine, 2s-2-tert-butoxycarbonyl amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, pubchem14939, n, a-boc-n, a-fmoc-l-lysine
PubChem CID 10983524
SMILES CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 93°C to 98°C
Color White
Beilstein 3576584
Solubility Information Soluble in dimethylformamide. Insoluble in water.
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-6-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Quantity 25 g
Odor Odorless
Formula Weight 468.55
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Nalpha-Boc-Nepsilon-Fmoc-L-lysine
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only