missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tryptophan 98.0+%, TCI America™
$61.61 - $246.43
Chemical Identifiers
| CAS | 35737-15-6 |
|---|---|
| Molecular Formula | C26H22N2O4 |
| Molecular Weight (g/mol) | 426.47 |
| MDL Number | MFCD00037126 |
| InChI Key | MGHMWKZOLAAOTD-DEOSSOPVSA-N |
| Synonym | fmoc-trp-oh, nalpha-fmoc-l-tryptophan, fmoc-l-tryptophan, n-fmoc-l-tryptophan, fmoc-l-trp-oh, fmoc-tryptophan, l-tryptophan, n-9h-fluoren-9-ylmethoxy carbonyl, n-9h-fluoren-9-ylmethoxy carbonyl-l-tryptophan, 9-fluorenylmethoxycarbonyl-l-tryptophan, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1h-indol-3-yl propanoic acid |
| PubChem CID | 978343 |
| IUPAC Name | (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(1H-indol-3-yl)propanoic acid |
| SMILES | OC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2 |
Chemical Identifiers
| 35737-15-6 | |
| 426.47 | |
| MGHMWKZOLAAOTD-DEOSSOPVSA-N | |
| 978343 | |
| OC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2 |
| C26H22N2O4 | |
| MFCD00037126 | |
| fmoc-trp-oh, nalpha-fmoc-l-tryptophan, fmoc-l-tryptophan, n-fmoc-l-tryptophan, fmoc-l-trp-oh, fmoc-tryptophan, l-tryptophan, n-9h-fluoren-9-ylmethoxy carbonyl, n-9h-fluoren-9-ylmethoxy carbonyl-l-tryptophan, 9-fluorenylmethoxycarbonyl-l-tryptophan, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1h-indol-3-yl propanoic acid | |
| (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(1H-indol-3-yl)propanoic acid |
Specifications
| 35737-15-6 | |
| White | |
| MFCD00037126 | |
| fmoc-trp-oh, nalpha-fmoc-l-tryptophan, fmoc-l-tryptophan, n-fmoc-l-tryptophan, fmoc-l-trp-oh, fmoc-tryptophan, l-tryptophan, n-9h-fluoren-9-ylmethoxy carbonyl, n-9h-fluoren-9-ylmethoxy carbonyl-l-tryptophan, 9-fluorenylmethoxycarbonyl-l-tryptophan, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1h-indol-3-yl propanoic acid | |
| OC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2 | |
| 426.47 | |
| 426.47 | |
| Crystalline Powder |
| 187°C | |
| C26H22N2O4 | |
| 5 g | |
| MGHMWKZOLAAOTD-DEOSSOPVSA-N | |
| (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(1H-indol-3-yl)propanoic acid | |
| 978343 | |
| ≥98.0% (HPLC,T) | |
| Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tryptophan |
Safety and Handling
TSCA : No