missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tryptophan 98.0+%, TCI America™

Supplier:  TCI America F030725G

 View more versions of this product

Catalog No. F030725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tryptophan
187°C
C26H22N2O4
25 g
MGHMWKZOLAAOTD-DEOSSOPVSA-N
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(1H-indol-3-yl)propanoic acid
978343
≥98.0% (HPLC,T)
35737-15-6
White
MFCD00037126
fmoc-trp-oh, nalpha-fmoc-l-tryptophan, fmoc-l-tryptophan, n-fmoc-l-tryptophan, fmoc-l-trp-oh, fmoc-tryptophan, l-tryptophan, n-9h-fluoren-9-ylmethoxy carbonyl, n-9h-fluoren-9-ylmethoxy carbonyl-l-tryptophan, 9-fluorenylmethoxycarbonyl-l-tryptophan, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1h-indol-3-yl propanoic acid
OC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
426.47
426.47
Crystalline Powder

Chemical Identifiers

35737-15-6
426.47
MGHMWKZOLAAOTD-DEOSSOPVSA-N
978343
OC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
C26H22N2O4
MFCD00037126
fmoc-trp-oh, nalpha-fmoc-l-tryptophan, fmoc-l-tryptophan, n-fmoc-l-tryptophan, fmoc-l-trp-oh, fmoc-tryptophan, l-tryptophan, n-9h-fluoren-9-ylmethoxy carbonyl, n-9h-fluoren-9-ylmethoxy carbonyl-l-tryptophan, 9-fluorenylmethoxycarbonyl-l-tryptophan, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1h-indol-3-yl propanoic acid
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(1H-indol-3-yl)propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions