accessibility menu, dialog, popup

UserName

CAS RN 35737-15-6

NH(S)OHNOHOO

CAS RN 35737-15-6

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(1H-indol-3-yl)propanoic acid
Synonyms: fmoc-tryptophan|FMOC-TRP-OH
Molecular Weight (g/mol): 426.47
Molecular Formula: C26H22N2O4
InChi Key: MGHMWKZOLAAOTD-DEOSSOPVSA-N
SMILES: OC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results