missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine tert-Butyl Ester 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 127°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 383.44 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine tert-Butyl Ester |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |