missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-tryptophan 97.0+%, TCI America™

Catalog No. F06095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06095G 5 g
F060925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06095G Supplier TCI America Supplier No. F06095G
Only null left

Chemical Identifiers

CAS 86123-11-7
Molecular Formula C26H21N2O4
Molecular Weight (g/mol) 425.47
MDL Number MFCD00062954
InChI Key MGHMWKZOLAAOTD-XMMPIXPASA-M
Synonym fmoc-d-trp-oh, fmoc-d-tryptophan, n-9-fluorenylmethoxycarbonyl-d-tryptophan, n-alpha-fmoc-d-tryptophan, fmoc-d-trp, n-a-fmoc-d-tryptophan, nalpha-9h-fluoren-9-ylmethoxy carbonyl-d-tryptophan, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-1h-indol-3-yl propanoic acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1h-indol-3-yl propanoic acid, d-tryptophan, n-9h-fluoren-9-ylmethoxy carbonyl
PubChem CID 978344
IUPAC Name (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(1H-indol-3-yl)propanoate
SMILES [O-]C(=O)[C@@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 184°C
Quantity 5 g
Formula Weight 426.47
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-tryptophan