missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-lysine Hydrochloride 98.0+%, TCI America™

Catalog No. F06045G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06045G 5 g
F06041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06045G Supplier TCI America Supplier No. F06045G
Only null left

Chemical Identifiers

CAS 201002-47-3
Molecular Formula C21H25ClN2O4
Molecular Weight (g/mol) 404.891
MDL Number MFCD00190888
InChI Key MVMZFAIUUXYFGY-FSRHSHDFSA-N
Synonym Nalpha-Fmoc-D-lysine Hydrochloride, Fmoc-D-Lys-OH.HCl
PubChem CID 44629939
IUPAC Name (2R)-6-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid;hydrochloride
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCCN)C(=O)O.Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 404.89
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-lysine Hydrochloride
TSCA No
Recommended Storage Refrigerator