missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalidixic Acid 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 229°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 232.24 |
| Percent Purity | ≥95.0% (T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Nalidixic Acid |
Safety and Handling
| RTECSNumber | QN2885000 |
| TSCA | No |