missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Wash Bottles

Description
High quality squeeze bottles with wide mouth for easy filling. Thermo Scientific™ Nalgene™ Wide Mouth LDPE Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.
- Leakproof† bottle comes with polypropylene copolymer draw tube and polypropylene screw closure.
- Tip can be cut back for increased flow.
- Tight fit of draw tube and closure prevents separation when squeezing bottle.
Specifications
Specifications
| Capacity | 16 oz., 500 mL |
| Capacity (Metric) | 500 mL |
| Color | Translucent |
| Description | Polypropylene copolymer draw tube |
| Material | LDPE |
| No. per Case | 24 |
| No. per Pack | 4 |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide |
| Show More |