Learn More
Thermo Scientific™ Nalgene™ Straight-Sided Wide-Mouth Polycarbonate Jars with Closure
GREENER_CHOICE
Collect and store pastes, solids, specimens and lab supplies in these clear, straight-sided wide-mouth PC jars, which are high impact-resistant and extremely versatile.
$64.20 - $350.14
Specifications
Color Clear
Material Polycarbonate
No. per Pack 4
Clarity Clear
Mouth Wide
View More Specs
Products 7
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
029251D
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21160015
15 mL 38 mm
Pack of 4 Each for $64.20
Case of 12 Pack for $770.40
Only null left
029251A
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21160030
30 mL 43 mm
Pack of 4 Each for $82.07
Case of 12 Pack for $984.84
Only null left
029251B
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21160060
60 mL 53 mm
Pack of 4 Each for $85.23
Case of 12 Pack for $1,022.76
Only null left
029251C
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21160125
125 mL 70 mm
Pack of 4 Each for $161.64
Case of 6 Pack for $969.84
Only null left
029251G
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21160250
250 mL 70 mm
Pack of 4 Each for $186.25
Case of 6 Pack for $1,117.50
Only null left
029251H
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21160500
500 mL 120 mm
Pack of 4 Each for $293.64
Case of 4 Pack for $1,174.56
Only null left
029251J
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21161000
1000 mL 120 mm
Pack of 4 Each for $350.14
Case of 4 Pack for $1,400.56
Only null left
Description
Description
Collect and store pastes, solids, specimens and lab supplies with window-clear Thermo Scientific™ Nalgene™ Straight-side Wide-Mouth Jars. A wide temperature range (−135°C to +135°C) makes these jars an excellent choice for cold room and refrigerated use all the way to the autoclave. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables. These high impact-resistant jars are extremely versatile. Not liquid leakproof.
  • Excellent for cold room and refrigerated use — easy to see contents
  • Wide-mouth and straight sides provide full-access and complete sample retrieval ease
  • 15, 30 and 60mL jars have natural polypropylene closures
  • 125mL to 1L jars have white polypropylene closures with recessed tops for stacking
  • 125 and 250mL sizes fit Osterizer™ and other blenders that have Mason Jar threads; recommended for use with Fluid Transfer Closure DS2153-0700
  • Not liquid leakproof; not recommended for liquid transport
  • Compatible with most weak acids, aliphatic alcohols and aliphatic hydrocarbons.  Not for use with solvents, bases, strong acids, and other harsh laboratory chemicals.  Consult the Nalgene Chemical Resistance guide for more detailed chemical compatibility ratings. 
  • Autoclavable/Transparent
Note: Completely disengage threads or remove cap before autoclaving.
Recycling Information:
Please recycle the corrugate cartons and LDPE bags in which your Nalgene  products are packaged.
Specifications
Specifications
Clear
4
Wide
Straight-sided Jar
Screw Cap
Polycarbonate
Clear
Round
Polypropylene