Learn More
Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays

Package and ship liquid medias, buffers and sera using these square, heavy-walled, durable bottles that offer excellent gas barrier properties.
$724.41 - $2,150.00
Specifications
| Color | Clear |
|---|---|
| For Use With (Application) | Storing Media and Sera |
| Material | PETG |
| Sterility | Non-sterile |
| Certifications/Compliance | USP Class VI, FDA Compliance 21 CFR |
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
0292382
![]() |
Thermo Scientific™
3220200030 |
30 mL | 20-415 mm |
Case of 280 Each for $2,150.00
|
|
||||
14178168
![]() |
Thermo Scientific™
3220200060 |
60 mL | 24-415 mm |
Case of 200 Each for $1,704.44
|
|
||||
0331334D
![]() |
Thermo Scientific™
3220200250 |
250 mL | 38-430 mm |
Case of 60 Each for $890.83
|
|
||||
0992315B
![]() |
Thermo Scientific™
3220200500 |
500 mL | 38-430 mm |
Case of 40 Each for $724.41
|
|
||||
0331334F
![]() |
Thermo Scientific™
3220201000 |
1 L | 38-430 mm |
Case of 24 Each for $858.31
|
|
||||
0331334G
![]() |
Thermo Scientific™
3220202000 |
2 L | 53B |
Case of 12 Each for $906.79
|
|
||||
Description
Package and ship liquid media, buffers and sera using Thermo Scientific™ Nalgene™ PETG Square Media Bottles with Closures. Heavy-walled, durable bottles offer excellent gas barrier properties and are specifically designed for storing media and sera.
- Heavy-walled and durable
- Shatter-resistant
- Provide excellent gas barrier properties
- In shrink-wrap trays
- Molded in graduations
- Clear bottles for easy identification of bottles
- Nonsterile
- Leakproof†
Specifications
| Clear | |
| PETG | |
| USP Class VI, FDA Compliance 21 CFR | |
| Yes | |
| Square | |
| HDPE |
| Storing Media and Sera | |
| Non-sterile | |
| Yes | |
| Narrow | |
| Media Bottle |