Learn More
Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays

Package and ship liquid medias, buffers and sera using these square, heavy-walled, durable bottles that offer excellent gas barrier properties.
Supplier: Thermo Scientific™ 3220200500
Description
Package and ship liquid media, buffers and sera using Thermo Scientific™ Nalgene™ PETG Square Media Bottles with Closures. Heavy-walled, durable bottles offer excellent gas barrier properties and are specifically designed for storing media and sera.
- Heavy-walled and durable
- Shatter-resistant
- Provide excellent gas barrier properties
- In shrink-wrap trays
- Molded in graduations
- Clear bottles for easy identification of contents
- Nonsterile
- Leakproof†
Note:
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Chemical Identifiers
| Clear | |
| PETG | |
| USP Class VI, FDA Compliance 21 CFR | |
| Yes | |
| Square | |
| HDPE |
| Storing Media and Sera | |
| Non-sterile | |
| Yes | |
| Narrow | |
| Media Bottle |
Specifications
| 500 mL | |
| Clear | |
| 2.9 in. | |
| 28 mm | |
| PETG | |
| 20 | |
| USP Class VI, FDA Compliance 21 CFR | |
| Yes | |
| Square | |
| Media Bottle | |
| HDPE | |
| 74 mm |
| 16.9 oz. | |
| 500mL, 38-430 closure, tray pack | |
| 1.1 in. | |
| Storing Media and Sera | |
| 40 | |
| Non-sterile | |
| Yes | |
| Narrow | |
| 177 mm | |
| 6.97 in. | |
| 38-430 mm |