missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays
GREENER_CHOICE

Catalog No. 0292382
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
250 mL
500 mL
1 L
2 L
Closure Size:
53B
20-415 mm
24-415 mm
38-430 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
0292382 30 mL 20-415 mm
14178168 60 mL 24-415 mm
0331334D 250 mL 38-430 mm
0992315B 500 mL 38-430 mm
0331334F 1 L 38-430 mm
0331334G 2 L 53B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 0292382 Supplier Thermo Scientific™ Supplier No. 3220200030

Package and ship liquid medias, buffers and sera using these square, heavy-walled, durable bottles that offer excellent gas barrier properties.

Package and ship liquid media, buffers and sera using Thermo Scientific™ Nalgene™ PETG Square Media Bottles with Closures. Heavy-walled, durable bottles offer excellent gas barrier properties and are specifically designed for storing media and sera.

  • Heavy-walled and durable
  • Shatter-resistant
  • Provide excellent gas barrier properties
  • In shrink-wrap trays
  • Molded in graduations
  • Clear bottles for easy identification of contents
  • Nonsterile
  • Leakproof†

Note:

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications

Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Clear
Description 30mL, 20-415 closure, tray pack
Diameter (English) 1.5 in.
Diameter (English) Inner Neck 0.55 in.
Diameter (Metric) Inner Neck 14 mm
For Use With (Application) Storing Media and Sera
Material PETG
No. per Case 280
No. per Pack 40
Sterility Non-sterile
Certifications/Compliance USP Class VI, FDA Compliance 21 CFR
Closure Included Yes
Graduated Yes
Mouth Narrow
Shape Square
Height (Metric) 64 mm
Product Type Media Bottle
Height (English) 2.5 in.
Closure Material HDPE
Closure Size 20-415 mm
Diameter (Metric) 38 mm
Show More Show Less