Learn More
Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom: Sterile
Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing, and storage.
$345.25 - $1190.06
Specifications
Material PETG
No. per Pack 1
Surface Coating None
Autoclavable Not Autoclavable
Disposable Single-use
View More Specs
Products 12
Catalog Number Mfr. No. Capacity (Metric) Closure Size Closure Type Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Closure Type Price Quantity  
10-041-23
View Documents
Thermo Scientific™
41130125
125 mL 38-430 mm Solid, non-vented
Case of 24 Each for $972.22
Only null left
10-346-8A
View Documents
Thermo Scientific™
41130250
250 mL 38-430 mm Solid, non-vented
Case of 12 Each for $601.53
Only null left
10-346-8B
View Documents
Thermo Scientific™
41130500
500 mL 45-430 mm Solid, non-vented
Case of 12 Each for $666.78
Only null left
10-041-24
View Documents
Thermo Scientific™
41131000
1000 mL 45-430 mm Solid, non-vented
Case of 6 Each for $526.75
Only null left
10-346-8C
View Documents
Thermo Scientific™
41132000
2000 mL 45-430 mm Solid, non-vented
Case of 4 Each for $445.47
Only null left
03-600-151
View Documents
Thermo Scientific™
41132800
2800 mL 70 mm Solid, non-vented
Case of 4 Each for $734.88
Only null left
10-529
View Documents
Thermo Scientific™
41160125
125 mL 38-430 mm Vented
Case of 24 Each for $1,190.06
Only null left
10-530
View Documents
Thermo Scientific™
41160250
250 mL 38-430 mm Vented
Case of 12 Each for $695.84
Only null left
10-531
View Documents
Thermo Scientific™
41160500
500 mL 45-430 mm Vented
Case of 12 Each for $851.91
Only null left
10-532
View Documents
Thermo Scientific™
41161000
1000 mL 45-430 mm Vented
Case of 6 Each for $540.41
Only null left
10-533
View Documents
Thermo Scientific™
41162000
2000 mL 45-430 mm Vented
Case of 4 Each for $345.25
Only null left
03-600-153
View Documents
Thermo Scientific™
41162800
2800 mL 70 mm Vented
Case of 4 Each for $735.31
Only null left
Description
Description
Save preparation time and avoid contamination risk by using Thermo Scientific Nalgene Single-Use PETG Erlenmeyer Flasks with Baffled Bottom, ideal for suspension of cell culture, media preparation, mixing, and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
Features
  • Sterile and single use to reduce contamination risk
  • BPA-free
  • Collapses when autoclaved to reduce bio hazardous waste
  • Graduated for quick volume assessment
Warranty: 90 days
Specifications
Specifications
PETG
None
Single-use
Sterile
1
Not Autoclavable
Yes
Baffled
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.