missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom: Sterile

Catalog No. 1004123
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
2000 mL
2800 mL
Closure Size:
70 mm
38-430 mm
45-430 mm
Closure Type:
Solid, non-vented
Vented
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size Closure Type
1004123 125 mL 38-430 mm Solid, non-vented
103468A 250 mL 38-430 mm Solid, non-vented
103468B 500 mL 45-430 mm Solid, non-vented
1004124 1000 mL 45-430 mm Solid, non-vented
103468C 2000 mL 45-430 mm Solid, non-vented
03600151 2800 mL 70 mm Solid, non-vented
10529 125 mL 38-430 mm Vented
10530 250 mL 38-430 mm Vented
10531 500 mL 45-430 mm Vented
10532 1000 mL 45-430 mm Vented
10533 2000 mL 45-430 mm Vented
03600153 2800 mL 70 mm Vented
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 1004123 Supplier Thermo Scientific™ Supplier No. 41130125

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing, and storage.

Save preparation time and avoid contamination risk by using Thermo Scientific Nalgene Single-Use PETG Erlenmeyer Flasks with Baffled Bottom, ideal for suspension of cell culture, media preparation, mixing, and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.

Features

  • Sterile and single use to reduce contamination risk
  • BPA-free
  • Collapses when autoclaved to reduce bio hazardous waste
  • Graduated for quick volume assessment

Warranty: 90 days

Specifications

Capacity (English) 4 oz.
Capacity (Metric) 125 mL
Material PETG
No. per Case 24
No. per Pack 1
Shape Erlenmeyer
Surface Coating None
Autoclavable Not Autoclavable
Disposable Single-use
Graduated Yes
Mouth Style Narrow
Sterility Sterile
Bottom Shape Baffled
Closure Type Solid, non-vented
Closure Size 38-430 mm
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.