Learn More
Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom: Sterile
Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing, and storage.
Supplier: Thermo Scientific™ 41160125
Description
Save preparation time and avoid contamination risk by using Thermo Scientific Nalgene Single-Use PETG Erlenmeyer Flasks with Baffled Bottom, ideal for suspension of cell culture, media preparation, mixing, and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
Features
- Sterile and single use to reduce contamination risk
- BPA-free
- Collapses when autoclaved to reduce bio hazardous waste
- Graduated for quick volume assessment
Warranty: 90 days
Specifications
| 4 oz. | |
| PETG | |
| 1 | |
| None | |
| Single-use | |
| Narrow | |
| Baffled | |
| 38-430 mm |
| 125 mL | |
| 24 | |
| Erlenmeyer | |
| Not Autoclavable | |
| Yes | |
| Sterile | |
| Vented |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.