Learn More
Thermo Scientific™ Nalgene™ Rectangular Polypropylene Tank with Cover
Significantly reduce evaporation and contamination by using these covered, rectangular tanks, which are offered in sizes from 8 to 114L to meet application needs.
Supplier:  Thermo Scientific™ 142000005
 Afficher plus de versions de ce produit
Numéro de catalogue. 14-831-111
Il en reste null
Description
Description
Significantly reduce evaporation and contamination by using a Thermo Scientific™ Nalgene™ Rectangular Polypropylene Tank with Cover, offered in sizes ranging from 8 to 114L. Tanks can be modified with a factory-installed spigot.
  • Suitable for use with many organic chemicals
  • Offers good stress-crack resistance
  • Autoclavable
Note:
Tanks used at elevated temperatures or with high specific-gravity liquids should be externally supported.
Spécifications
Spécifications
14 in.
Polypropylene
20 L
10 in.
10 in.
Tank with Cover
5 gal.
35.6 cm
25 cm
25 cm
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Caution:
Plastic tanks are generally subject to more severe conditions that plastic labware; exposure is constant, stresses are greater and different classes and concentrations of chemicals are involved. Please pay special attention to chemical compatibility.