Learn More
Thermo Scientific™ Nalgene™ Rectangular HDPE Bottles with Closure: Bulk Pack
GREENER_CHOICE
Save space with the rectangular design of these rectangular HDPE bottles with closure, a good choice when space is at a premium.
$1,781.66 - $2,496.62
Specifications
Color Natural
Description Rectangular Bottle
Material HDPE
Sterility Non-sterile
Closure Color Natural
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
0331341B
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3120070008
250 mL 38 mm
Case of 250 Each for $2,496.62
Only null left
0331341C
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3120070016
500 mL 48 mm
Case of 125 Each for $1,931.24
Only null left
0331341D
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3120070032
1 L 53 mm
Case of 50 Each for $1,781.66
Only null left
Description
Description
Save space with the rectangular design of Thermo Scientific™ Nalgene™ Rectangular HDPE Bottles with Closure, a good choice when space is at a premium.
Features:
  • Resistant to splitting or puncturing
  • Constructed of high density polyethylene with polypropylene closure
  • Leakproof† when used with Nalgene closures
  • Bulk packed for packaging applications
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton.
Specifications
Specifications
Natural
HDPE
Natural
Yes
Wide
Bottle
Screw Cap
Rectangular Bottle
Non-sterile
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
No
Rectangular
Polypropylene
No
Videos
SDS
Documents
Documents
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.