missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene, Quick Action Spigot
GREENER_CHOICE

Catalog No. 029637
Change view
Click to view available options
Polypropylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
029637 Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 029637 Supplier Thermo Scientific™ Supplier No. 64220010
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Use With: Nalgene carboys

Stop drips and keep your Nalgene carboys in service for years with these Thermo Scientific™ Nalgene™ Polypropylene Quick-Action Replacement Spigots. Includes an HDPE tubing adapter which accepts tubing with 1/4 or 5/16 in. I.D for added versatility.

Features:

  • Spigot without tubing adapter accepts 5/8″ (1.6cm) I.D. tubing
  • Supplied with HDPE adapter for use with 1/4" or 5/16″ (0.6 or 0.8cm) tubing

Notes:

  • Disassemble spigot before autoclaving; tubing adapter is not autoclavable

Specifications

Description Quick-Action Spigot PP fits 1-1/8 in. spigot boss for carboys 1-1/8 inch
Diameter (English) Inner Tubing 5/8 in. (includes adapter for 1/4 or 5/16 in. tubing)
Diameter (Metric) Inner Tubing 1.6 cm (includes adapter for 0.6 or 0.8 cm tubing)
For Use With (Equipment) Nalgene Carboys
Material Polypropylene
No. per Pack 1 Ea.
Product Type Spigot