missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Pour Spout
GREENER_CHOICE

Catalog No. 0292314AA
Change view
Click to view available options
Thread Size (Metric):
S2 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
0292314AA S2 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0292314AA Supplier Thermo Scientific™ Supplier No. DS21760384

Enjoy easy one-handed liquid dispensing without leaving bottles completely open to the environment. Vented to allow smooth flow for easy liquid dispensing.

Enjoy easy one-handed liquid dispensing without leaving bottles completely open to the environment. Thermo Scientific™ Nalgene™ Polypropylene Pour Spout is made to fit all Nalgene bottles with 38-430 neck finish. Vented to allow smooth flow for easy liquid dispensing.

  • Used for all Nalgene bottles with 38-430 neck finish
  • Allows easy one-handed liquid dispensing without leaving bottles completely open to the environment

Specifications

Color Translucent
Description Pour Spout
Material Polypropylene
No. per Case 6
No. per Pack 6
Thread Size (Metric) S2 mm
Liner Material Polyethylene
Neck Finish S2 mm
For Use With (Equipment) Nalgene bottles with 38-430mm neck finish
Product Type Pouring Spout