Learn More
Thermo Scientific™ Nalgene™ Polypropylene, Closure with Barbed Bulkhead Fittings, 1/4 inch
GREENER_CHOICE
Catalog No. 0292314X
Change view
Click to view available options
Quantity:
Case of 4
Catalog No. Quantity
0292314X Case of 4
1 options
Catalog No. 0292314X Supplier Thermo Scientific™ Supplier No. DS21670002
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For Use With: Nalgene culture vessels
Use Thermo Scientific™ Nalgene™ Closures with Barbed Bulkhead Fittings to customize most Nalgene bottle and carboy closures for aseptic transfer of liquids.
  • Size 38mm-430 closures are predrilled allowing for easy, safe in-house assembly
  • Barbed at both ends to accept tubing inside and outside the container
  • Molded of white, autoclavable polypropylene, ready for in-house sterilization
Includes:
Instructions, drilling template and a set of two panel-mounted barbed fittings, acetal nuts and silicone gaskets.
Ordering Information:
Barbed bulkhead fittings also sold separately (6149-0001 and 6149-0002).
Specifications
Closure Size 38-430
Description Closure with Barbed Bulkhead Fitting
Diameter (English) Inner Tubing 1/4 in.
For Use With (Equipment) Nalgene culture vessels
Material Polypropylene
No. per Case 4/Cs.
No. per Pack 4 Pk.
Product Type Closure
Quantity Case of 4