Learn More
Thermo Scientific™ Nalgene™ PMP Griffin Low-Form Plastic Beakers
Continuously use these transparent, PMP beakers with easy-to-read, silk-screened graduation at temperatures of 150°C with intermittent service to 175°C.
$202.14 - $423.21
Specifications
| Material | PMP |
|---|---|
| Type | Beaker |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Graduated | Yes |
Alert:
Not for use on hot plates.
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|
02-591-15B
|
Thermo Scientific™
12030050 |
50 mL |
Pack of 12 Each for $266.07
Case of 3 Pack for $798.21
|
|
|||||
|
02-591-15C
|
Thermo Scientific™
12030100 |
100 mL |
Pack of 12 Each for $296.07
Case of 3 Pack for $888.21
|
|
|||||
|
02-591-15D
|
Thermo Scientific™
12030150 |
150 mL |
Pack of 12 Each for $423.21
Case of 3 Pack for $1,269.63
|
|
|||||
|
02-591-15E
|
Thermo Scientific™
12030250 |
250 mL |
Pack of 6 Each for $254.50
Case of 4 Pack for $1,018.00
|
|
|||||
|
02-591-15F
|
Thermo Scientific™
12030400 |
400 mL |
Pack of 6 Each for $388.79
Case of 4 Pack for $1,555.16
|
|
|||||
|
02-591-15G
|
Thermo Scientific™
12030600 |
600 mL |
Pack of 4 Each for $309.86
Case of 3 Pack for $929.58
|
|
|||||
|
02-591-15H
|
Thermo Scientific™
12031000 |
1000 mL |
Pack of 3 Each for $280.32
Case of 4 Pack for $1,121.28
|
|
|||||
|
02-591-15J
|
Thermo Scientific™
12032000 |
2000 mL |
Each for $202.14
Case of 4 Each for $808.56
|
|
|||||
|
02-591-15AA
|
Thermo Scientific™
12034000 |
4000 mL |
Each for $312.39
Case of 4 Each for $1,249.56
|
|
|||||
Description
Continuously use Thermo Scientific™ Nalgene™ PMP Griffin Low-Form Beakers at temperatures of 150°C with intermittent service to 175°C. These polymethylpentene beakers have the transparency of glass, excellent chemical resistance and heat tolerance. All sizes of beakers have easy-to-read, silk-screened graduations.
- Transparent PMP material allows for a clear view of beaker contents
- No-drip, single-spout design provides precision pouring control
- With ribs for easy stacking
- Silk screened with graduations, size code, resin code, catalog number, max. temperature and “No Flame” symbol for easy identification and safe use
- Autoclavable/Graduated/Transparent
- Meets ISO/DIN 7056 International Standards for Laboratory Plasticware Accuracy
Specifications
| PMP | |
| Autoclavable | |
| Yes | |
| Griffin, Low Form |
| Beaker | |
| Reusable | |
| ISO/DIN 7056 |