missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures
GREENER_CHOICE

Catalog No. 0292314C
Change view
Click to view available options
Thread Size (Metric):
20 mm
24 mm
28 mm
38 mm
43 mm
53 mm
63 mm
Product Type:
Replacement Cap
Spout Cap
Color:
Amber
Natural
White
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric) Product Type Color
0292314C 28 mm Replacement Cap Natural
02925113 20 mm Replacement Cap White
0292314B 24 mm Replacement Cap Natural
0292314M 38 mm Replacement Cap Natural
0292314DD 43 mm Replacement Cap Natural
0292316J 53 mm Replacement Cap Amber
0292313T 53 mm Replacement Cap Natural
0292314E 53 mm Replacement Cap Natural
0292314GG 63 mm Replacement Cap Natural
02945131 24 mm Spout Cap Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 0292314C Supplier Thermo Scientific™ Supplier No. 7121500280

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fit these replacement closures onto Thermo Scientific™ Nalgene™ narrow-mouth and wide-mouth bottles.

Fit Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures onto Nalgene narrow-mouth and wide-mouth bottles.

  • Made to fit Nalgene narrow-mouth and wide-mouth bottles

Specifications

Color Natural
Description Bottle Replacement Closures
Material Polypropylene
No. per Case 12
Thread Size (Metric) 28 mm
Mouth Narrow
For Use With (Equipment) Nalgene narrow-mouth and wide-mouth bottles
Shape Round
Product Type Replacement Cap

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.