Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles
Meet regulatory labeling requirements for chemical containers. These Right-to-Know Wash Bottles use standard, instantly recognizable symbols and colors.
$196.79 - $645.34
Specifications
| Description | RTK Wash Bottle |
|---|---|
| Material | LDPE |
| Certifications/Compliance | Meet requirements of OSHA Hazard Communication Regulation [29 CFR 1910.1200(f)(4)]. |
| Closure Included | Yes |
| Graduated | No |
| Catalog Number | Mfr. No. | Capacity (Metric) | Chemical Label | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Chemical Label | Price | Quantity & Availability | ||||
0340911B
![]() |
Thermo Scientific™
24250502 |
500 mL | Ethyl Alcohol |
Pack of 6 Each for $225.00
Case of 4 Pack for $900.00
|
|
||||
0340911C
![]() |
Thermo Scientific™
24250503 |
500 mL | Methanol |
Pack of 6 Each for $225.00
Case of 4 Pack for $900.00
|
|
||||
034099A
![]() |
Thermo Scientific™
24250506 |
500 mL | Sodium Hypochlorite |
Pack of 6 Each for $225.00
Case of 4 Pack for $900.00
|
|
||||
0340911D
![]() |
Thermo Scientific™
24250504 |
500 mL | Isopropanol |
Pack of 6 Each for $225.00
Case of 4 Pack for $900.00
|
|
||||
0340911E
![]() |
Thermo Scientific™
24250505 |
500 mL | Distilled Water |
Pack of 6 Each for $225.00
Case of 4 Pack for $900.00
|
|
||||
0340911L
![]() |
Thermo Scientific™
24250500 |
500 mL | Assorted |
Pack of 5 Each for $196.79
Case of 4 Pack for $787.16
|
|
||||
0340911G
![]() |
Thermo Scientific™
24251002 |
1000 mL | Ethyl Alcohol |
Pack of 4 Each for $215.10
Case of 3 Pack for $645.30
|
|
||||
0340911H
![]() |
Thermo Scientific™
24251003 |
1000 mL | Methanol |
Case of 12 Each for $645.34
|
|
||||
0340911J
![]() |
Thermo Scientific™
24251004 |
1000 mL | Isopropanol |
Pack of 4 Each for $215.10
Case of 3 Pack for $645.30
|
|
||||
0340911K
![]() |
Thermo Scientific™
24251005 |
1000 mL | Distilled Water |
Pack of 4 Each for $196.79
Case of 3 Pack for $590.37
|
|
||||
Description
Meet NFPA and ICS labeling standards for chemical containers with Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles, which use standard, instantly recognizable colors and GHS symbols. These low-density polyethylene bottles are flexible and easy to squeeze for ergonomic dispensing. Made from high quality laboratory grade plastic materials for dependably low leachables and extractables.
Features:
- Polypropylene or high-density polyethylene closure; polypropylene copolymer fill tube
- Self-venting, so contents stay in bottle; prevents self-dispensing
- Chemical name, International Chemical Society (ICS) chemical formula and Chemical Abstract Service (CAS) number printed on bottle for easy identification
- Color bar that corresponds to colored closure to prevent cross-contamination
- Designed to dispense through the top with a closure and stem molded in one piece for uniform service
- Tip that can be cut back to increase flow
- Tight fit of draw tube and closure that prevents separation when squeezing bottle
Ordering Information:
Available in packs of the same 16 or 32 oz. (500 or 1000mL) bottle, or in a five-bottle (16 oz./500mL only) assortment pack consisting of one each acetone, ethyl alcohol, isopropanol, methanol and distilled water.
.
Specifications
| RTK Wash Bottle | |
| Meet requirements of OSHA Hazard Communication Regulation [29 CFR 1910.1200(f)(4)]. | |
| No | |
| Cylindrical | |
| Screw Cap |
| LDPE | |
| Yes | |
| Narrow | |
| Safety Wash Bottle | |
| Yes |