Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles

Meet regulatory labeling requirements for chemical containers. These Right-to-Know Wash Bottles use standard, instantly recognizable symbols and colors.
Supplier: Thermo Scientific™ 24250505
Description
Meet NFPA and ICS labeling standards for chemical containers with Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles, which use standard, instantly recognizable colors and GHS symbols. These low-density polyethylene bottles are flexible and easy to squeeze for ergonomic dispensing. Made from high quality laboratory grade plastic materials for dependably low leachables and extractables.
Features:
- Polypropylene or high-density polyethylene closure; polypropylene copolymer fill tube
- Self-venting, but contents stay in bottle with no risk of leakage
- Chemical name, International Chemical Society (ICS) chemical formula and Chemical Abstract Service (CAS) number printed on bottle for easy identification
- Color bar that corresponds to colored closure to prevent cross-contamination
- Designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service
- Tip that can be cut back to increase flow
- Tight fit of draw tube and closure that prevents separation when squeezing bottle
Ordering Information:
Available in packs of the same 16 or 32 oz. (500 or 1000mL) bottle, or in a five-bottle (16 oz./500mL only) assortment pack consisting of one each acetone, ethyl alcohol, isopropanol, methanol and distilled water.
Chemical Identifiers
| RTK Wash Bottle | |
| Meet requirements of OSHA Hazard Communication Regulation [29 CFR 1910.1200(f)(4)]. | |
| No | |
| Cylindrical | |
| Screw Cap |
| LDPE | |
| Yes | |
| Narrow | |
| Safety Wash Bottle | |
| Yes |
Specifications
| 16 oz., 500 mL | |
| 16 oz. | |
| RTK Wash Bottle | |
| 24 | |
| Natural | |
| Yes | |
| Narrow | |
| 210.82 mm | |
| 8.3 in. | |
| Polypropylene | |
| 72.64 mm | |
| Yes |
| 500 mL | |
| Natural | |
| LDPE | |
| 6 | |
| Meet requirements of OSHA Hazard Communication Regulation [29 CFR 1910.1200(f)(4)]. | |
| No | |
| Cylindrical | |
| Safety Wash Bottle | |
| Distilled Water | |
| 28 mm | |
| Screw Cap | |
| Not Autoclavable |