missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles
GREENER_CHOICE

Catalog No. 034099A
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Acetone
Assorted
Distilled Water
Ethyl Alcohol
Isopropanol
Methanol
Sodium Hypochlorite
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Chemical Label
034099A 500 mL Sodium Hypochlorite
0340911B 500 mL Ethyl Alcohol
0340911C 500 mL Methanol
0340911D 500 mL Isopropanol
0340911E 500 mL Distilled Water
0340911L 500 mL Assorted
0340911A 500 mL Acetone
0340911G 1000 mL Ethyl Alcohol
0340911H 1000 mL Methanol
0340911J 1000 mL Isopropanol
0340911K 1000 mL Distilled Water
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 034099A Supplier Thermo Scientific™ Supplier No. 24250506
Get a Water Bottle with Plastics Purchase
View all promotions

missing translation for 'validMainframeMsg'

Meet regulatory labeling requirements for chemical containers. These Right-to-Know Wash Bottles use standard, instantly recognizable symbols and colors.

Meet NFPA and ICS labeling standards for chemical containers with Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles, which use standard, instantly recognizable colors and GHS symbols. These low-density polyethylene bottles are flexible and easy to squeeze for ergonomic dispensing. Made from high quality laboratory grade plastic materials for dependably low leachables and extractables.

Features:

  • Polypropylene or high-density polyethylene closure; polypropylene copolymer fill tube
  • Self-venting, but contents stay in bottle with no risk of leakage
  • Chemical name, International Chemical Society (ICS) chemical formula and Chemical Abstract Service (CAS) number printed on bottle for easy identification
  • Color bar that corresponds to colored closure to prevent cross-contamination
  • Designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service
  • Tip that can be cut back to increase flow
  • Tight fit of draw tube and closure that prevents separation when squeezing bottle

Ordering Information:

Available in packs of the same 16 or 32 oz. (500 or 1000mL) bottle, or in a five-bottle (16 oz./500mL only) assortment pack consisting of one each acetone, ethyl alcohol, isopropanol, methanol and distilled water.

Specifications

Capacity 16 oz., 500 mL
Capacity (Metric) 500 mL
Color White
Description RTK Wash Bottle
Material LDPE
No. per Case 24
No. per Pack 6
Closure Color White
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 210.82 mm
Height (English) 8.3 in.
Chemical Label Sodium Hypochlorite
Closure Material HDPE
Closure Size 28 mm
Diameter (Metric) 72.64 mm
Closure Type Screw Cap
Through the Closure Dispensing Yes
Autoclavable Not Autoclavable
Capacity (English) 16 oz.
Shape Cylindrical
Certifications/Compliance Meet requirements of OSHA Hazard Communication Regulation [29 CFR 1910.1200(f)(4)].
Product Type Safety Wash Bottle
Show More Show Less