missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Dropper Bottles
GREENER_CHOICE

Catalog No. 0300719C
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
Closure Color:
Assorted
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Color
0300719C 15 mL Assorted
0300717A 4 mL White
0300717B 8 mL White
0300717C 15 mL White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 0300719C Supplier Thermo Scientific™ Supplier No. 27539050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use for accurate, reliable, repeatable dispensing of various reagents. These dropper bottles are made of opaque LDPE and are ideal for UV lightsensitive solutions. An excellent alternative to pipetting devices.

Choose Thermo Scientific™ Nalgene™ LDPE White Dropper Bottles for accurate, reliable, repeatable dispensing of various reagents and as an excellent alternative to pipetting devices. Nalgene™ Dropper Bottles are made of opaque LDPE and are ideal for UV lightsensitive solutions. Delivers 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time. Excellent chemical resistance; suitable for most biotech, diagnostic and pharmaceutical applications.

  • Dropper control tip snaps into place for a secure fit
  • Dispensing tip and closures included, choose from white or multi-colored caps
  • Leakproof†

Specifications

Capacity (Metric) 15 mL
Color White
Description LDPE Dropper Bottle
Includes Dispensing tip and closures
Material LDPE
No. per Case 25
No. per Pack 25
Closure Color Assorted
Closure Included Yes
Drop Size 40 μL
For Use With (Equipment) Nalgene Closures
Closure Size 15-415 mm
Closure Type Dropper Control Tip
Through the Closure Dispensing Yes
Capacity (English) 0.5 oz.
Shape Round
Product Type Dropping Bottle
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.