missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners
GREENER_CHOICE

Catalog No. 02540619
Change view
Click to view available options
Capacity (English):
5 gal.
7.5 gal.
10 gal.
15 gal.
30 gal.
55 gal.
80 gal.
100 gal.
150 gal.
200 gal.
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
02540619 200 gal.
02540610 5 gal.
02540611 7.5 gal.
02540612 10 gal.
02540613 15 gal.
02540614 30 gal.
02540615 55 gal.
02540616 80 gal.
02540617 100 gal.
02540618 150 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 02540619 Supplier Thermo Scientific™ Supplier No. 3430500200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Enhance mixing capabilities open-bag, flat-bottom Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners , which offer cost-effective, single-use fluid processing.

Enhance mixing capabilities with open-bag, flat-bottom liners. Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners offer cost-effective, single-use biopharmaceutical and diagnostic reagent fluid processing with rigid Nalgene cylindrical plastic tanks.

  • Designed to fit Nalgene cylindrical tanks from 5 gal (19L) to 200 gal (787L)
  • Gamma-irradiated, food-grade film is noncytotoxic and certified nonpyrogenic

Specifications

Type Tank Liner
Material Polyethylene
Capacity (English) 200 gal.
Capacity (Metric) 757 L
For Use With (Equipment) Nalgene cylindrical tanks from 5 gal (19L) to 200
No. per Case 10
No. per Pack 10

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.